
4-Fluoro-2-nitrophenyl trifluoromethanesulphonate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:722536-28-9
:
Formula:C₇H₃F₄NO₅S
MDL Number:MFCD25541894
:289.161 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together