
Methyl 2-(bromomethyl)-5-(trifluoromethyl)benzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:875895-66-2
:
Formula:C₁₀H₈BrF₃O₂
MDL Number:MFCD27931130
:297.069 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together