
Methyl 4-bromo-2-(trifluoromethyl)benzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:957207-58-8
:
Formula:C₉H₆BrF₃O₂
MDL Number:MFCD12761599
:283.042 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together