
2-Nitro-4-(trifluoromethoxy)phenyl trifluoromethanesulphonate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:955395-91-2
:
Formula:C₈H₃F₆NO₆S
MDL Number:MFCD25541876
ADR:8,III
:355.168 g/mol
UN:3265
:GHS05
Storage Temperature:Ambient
Frequently Bought Together