Order Entry
Netherlands
ContactUsLinkComponent
Methyl 4-bromo-2-chloro-6-fluorobenzoate 97%
Methyl 4-bromo-2-chloro-6-fluorobenzoate 97%
Catalog # APOSPC49657-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 4-bromo-2-chloro-6-fluorobenzoate 97%
Catalog # APOSPC49657-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :267.48 g/mol
Storage Temperature:Ambient
CAS Number:1321613-02-8
Formula:C₈H₅BrClFO₂
MDL Number:MFCD22574196