
2-Bromo-4,5-difluorobenzyl alcohol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:476620-55-0
:
Formula:C₇H₅BrF₂O
MDL Number:MFCD18909667
:223.015 g/mol
Melting Pt:75…76 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together