
4-(Aminomethyl)-2-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:916304-20-6
:
Formula:C₇H₇F₃N₂
MDL Number:MFCD11100735
:176.139 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together