
4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}-3-hydroxybutanenitrile 97%
Catalog # APOSPC4919-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:279.646 g/mol
Melting Pt:107…109 °C
:GHS07
Storage Temperature:Ambient
CAS Number:338966-48-6
Formula:C₁₀H₉ClF₃N₃O
MDL Number:MFCD01315213