
Methyl 4-bromo-2-(trifluoromethoxy)benzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:933785-18-3
:
Formula:C₉H₆BrF₃O₃
MDL Number:MFCD12761592
:299.041 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together