
4-(Dimethylsulphamoyl)-3-fluorobenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1534724-80-5
:
Formula:C₉H₁₀FNO₄S
MDL Number:MFCD21817646
:247.243 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together