Dipotassium 1,1,1,4,4,4-hexafluoro-2,3-bis(trifluoromethyl)butane-2,3-bis(olate) 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:127802-89-5
:
Formula:C₆F₁₂O₂·2K
MDL Number:MFCD27918470
:410.24 g/mol
Storage Temperature:Ambient
Frequently Bought Together