Methyl perfluoropentyl ether
Catalog # APOSPC450088-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:300.07 g/mol
Storage Temperature:Ambient
CAS Number:181214-74-4
Formula:C₆H₃F₁₁O
MDL Number:MFCD16620574