Order Entry
Netherlands
ContactUsLinkComponent
[(1R)-1-(2-Fluorophenyl)ethyl][(1S)-1-phenylethyl]ammonium fumarate
[(1R)-1-(2-Fluorophenyl)ethyl][(1S)-1-phenylethyl]ammonium fumarate
Catalog # APOSPC450077-250MG
Supplier:  Apollo Scientific
CAS Number:  
[(1R)-1-(2-Fluorophenyl)ethyl][(1S)-1-phenylethyl]ammonium fumarate
Catalog # APOSPC450077-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :359.391 g/mol
Storage Temperature:Ambient
Formula:C₁₆H₁₈FN·C₄H₄O₄
MDL Number:MFCD25978112