
4-(2-Fluoroethyl)piperidine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:184044-25-5
:
Formula:C₇H₁₄FN·ClH
MDL Number:MFCD27918502
:167.652 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together