Order Entry
Netherlands
ContactUsLinkComponent
(1S)-1-(3,5-Difluorophenyl)-N-[(1S)-1-phenylethyl]ethanaminium 2-carboxybenzoate
(1S)-1-(3,5-Difluorophenyl)-N-[(1S)-1-phenylethyl]ethanaminium 2-carboxybenzoate
Catalog # APOSPC450029-250MG
Supplier:  Apollo Scientific
CAS Number:  
(1S)-1-(3,5-Difluorophenyl)-N-[(1S)-1-phenylethyl]ethanaminium 2-carboxybenzoate
Catalog # APOSPC450029-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :427.441 g/mol
Storage Temperature:Ambient
CAS Number:444643-21-4
Formula:C₁₆H₁₈F₂N·C₈H₅O₄
MDL Number:MFCD28397825