
2-(1,1,2,2-Tetrafluoro-2-phenylethyl)pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1378261-29-0
:
Formula:C₁₃H₉F₄N
MDL Number:MFCD22377940
:255.211 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together