[1-(4-Trifluoromethylphenyl)cyclopropyl]methanol
Catalog # APOSPC449013-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:216.2 g/mol
Storage Temperature:Ambient
CAS Number:850406-46-1
Formula:C₁₁H₁₁F₃O
MDL Number:MFCD07374457