6-Fluoro-3-methylbenzo[d]isoxazol-5-amine hydrochloride
Catalog # APOSPC448017-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:202.613 g/mol
Storage Temperature:Ambient
CAS Number:1243313-45-2
Formula:C₈H₇FN₂O·ClH
MDL Number:MFCD16036053