1-(3-Trifluoromethyl-benzyl)-1H-pyrazole-4-carboxylic acid ethyl ester
Catalog # APOSPC446073-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:298.26 g/mol
Storage Temperature:Ambient
CAS Number:1035224-53-3
Formula:C₁₄H₁₃F₃N₂O₂
MDL Number:MFCD24386141