1-Phenyl-3-trifluoromethyl-1H-pyrazole-4-carboxylic acid
Catalog # APOSPC446065-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:256.181 g/mol
Storage Temperature:Ambient
CAS Number:142818-01-7
Formula:C₁₁H₇F₃N₂O₂
MDL Number:MFCD11707231