Sodium (E)-1-cyano-1-ethoxycarbonyl-3,3,3-trifluoro-propen-2-olate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1311283-96-1
:
Formula:C₇H₅F₃NO₃·Na
MDL Number:MFCD19981460
:231.104 g/mol
Storage Temperature:Ambient
Frequently Bought Together