4-Chloro-N-(6-chloro-4-trifluoromethyl-pyridin-2-yl)-benzamide
Catalog # APOSPC446046-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:335.109 g/mol
Storage Temperature:Ambient
CAS Number:1311278-37-1
Formula:C₁₃H₇Cl₂F₃N₂O
MDL Number:MFCD19981226