6-Fluoro-2-(pyrrolidin-3-yl)isoindolin-1-one
Catalog # APOSPC430581-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:220.243 g/mol
Storage Temperature:Ambient
CAS Number:1206969-88-1
Formula:C₁₂H₁₃FN₂O
MDL Number:MFCD14585250