Order Entry
Netherlands
ContactUsLinkComponent
N-(4-Fluorobenzyl)-6-chloropyrazin-2-amine
N-(4-Fluorobenzyl)-6-chloropyrazin-2-amine
Catalog # APOSPC430566-250MG
Supplier:  Apollo Scientific
CAS Number:  
N-(4-Fluorobenzyl)-6-chloropyrazin-2-amine
Catalog # APOSPC430566-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :237.661 g/mol
Storage Temperature:Ambient
CAS Number:629670-16-2
Formula:C₁₁H₉ClFN₃
MDL Number:MFCD11790282