Order Entry
Netherlands
ContactUsLinkComponent
(3,9-Diaza-spiro[5.5]undec-3-yl)(2-fluoro-phenyl)methanone
(3,9-Diaza-spiro[5.5]undec-3-yl)(2-fluoro-phenyl)methanone
Catalog # APOSPC430549-100MG
Supplier:  Apollo Scientific
CAS Number:  
(3,9-Diaza-spiro[5.5]undec-3-yl)(2-fluoro-phenyl)methanone
Catalog # APOSPC430549-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :276.349 g/mol
Storage Temperature:Ambient
CAS Number:1206970-65-1
Formula:C₁₆H₂₁FN₂O
MDL Number:MFCD14585279