N-(4-Fluorobenzyl)-6-chloropyridazin-3-amine
Catalog # APOSPC430543-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:237.661 g/mol
Storage Temperature:Ambient
CAS Number:872102-12-0
Formula:C₁₁H₉ClFN₃
MDL Number:MFCD07681959