1-N-Boc-4-Fluoropiperidine
Catalog # APOSPC430322-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:203.254 g/mol
Storage Temperature:Ambient
CAS Number:178181-55-0
Formula:C₁₀H₁₈FNO₂
MDL Number:MFCD11054109