Order Entry
Netherlands
ContactUsLinkComponent
2-(2-Fluorophenyl)-3H-imidazo[4,5-c]pyridine methanesulphonate
2-(2-Fluorophenyl)-3H-imidazo[4,5-c]pyridine methanesulphonate
Catalog # APOSPC430102-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(2-Fluorophenyl)-3H-imidazo[4,5-c]pyridine methanesulphonate
Catalog # APOSPC430102-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :309.316 g/mol
Storage Temperature:Ambient
CAS Number:1221449-52-0
Formula:C₁₂H₈FN₃·CH₃SO₃H
MDL Number:MFCD16040217