2,5-Difluoro-1a,7,7a,7b-tetrahydro-2H-oxireno[2,3-c]chromene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:944950-67-8
:
Formula:C₉H₈F₂O₂
MDL Number:MFCD09953723
:186.155 g/mol
Storage Temperature:Ambient
Frequently Bought Together