2-Ethoxy-6-trifluoromethylpyridine-4-boronic acid, pinacol ester 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1256359-30-4
:
Formula:C₁₄H₁₉BF₃NO₃
MDL Number:MFCD17214287
:317.112 g/mol
Storage Temperature:Ambient
Frequently Bought Together