Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Trifluoromethyl-1H-pyrazol-4-ylboronic acid
5-Trifluoromethyl-1H-pyrazol-4-ylboronic acid 97%
5-Trifluoromethyl-1H-pyrazol-4-ylboronic acid 97%
APOSPC412168-1G
:
Apollo Scientific
:
5-Trifluoromethyl-1H-pyrazol-4-ylboronic acid 97%
APOSPC412168-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
179.893 g/mol
:
GHS07
:
Freezer
:
1202054-12-3
:
C₄H₄BF₃N₂O₂
:
MFCD07375369