7-Fluoro-5-azaspiro[2.4]heptane hydrochloride
Catalog # APOSPC411283-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:151.61 g/mol
Storage Temperature:Ambient
CAS Number:2227205-26-5
Formula:C₆H₁₀FN·ClH
MDL Number:MFCD31697808