3-(Trifluoromethoxy)azetidine hydrochloride
Catalog # APOSPC411266-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:177.553 g/mol
Storage Temperature:Ambient
CAS Number:1803590-72-8
Formula:C₄H₆F₃NO·ClH
MDL Number:MFCD26097230