Order Entry
Netherlands
ContactUsLinkComponent
4-Chloro-1-[(3-fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid
4-Chloro-1-[(3-fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid
Catalog # APOSPC410499-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-1-[(3-fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid
Catalog # APOSPC410499-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :270.644 g/mol
Storage Temperature:Ambient
CAS Number:1006489-14-0
Formula:C₁₁H₈ClFN₂O₃
MDL Number:MFCD04968048