Order Entry
Netherlands
ContactUsLinkComponent
 
1-(2,2-Difluoroethyl)-3-(trifluoromethyl)-1H-pyrazole-5-carboxylic acid
Catalog # APOSPC410463-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2,2-Difluoroethyl)-3-(trifluoromethyl)-1H-pyrazole-5-carboxylic acid
Catalog # APOSPC410463-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :244.119 g/mol
Storage Temperature:Ambient
CAS Number:1006462-23-2
Formula:C₇H₅F₅N₂O₂
MDL Number:MFCD08696530