Order Entry
Netherlands
ContactUsLinkComponent
3-[1'-Methyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propanenitrile
3-[1'-Methyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propanenitrile
Catalog # APOSPC410462-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[1'-Methyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propanenitrile
Catalog # APOSPC410462-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :269.226 g/mol
Storage Temperature:Ambient
CAS Number:1006461-64-8
Formula:C₁₁H₁₀F₃N₅
MDL Number:MFCD08696393