Order Entry
Netherlands
ContactUsLinkComponent
3-[3-Methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
3-[3-Methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
Catalog # APOSPC410461-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[3-Methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
Catalog # APOSPC410461-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :203.164 g/mol
Storage Temperature:Ambient
CAS Number:1006461-11-5
Formula:C₈H₈F₃N₃
MDL Number:MFCD08558151