N-[(1-Methyl-1H-pyrazol-3-yl)methyl]-2-(trifluoromethyl)aniline
Catalog # APOSPC410454-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:255.239 g/mol
Storage Temperature:Ambient
CAS Number:1006449-98-4
Formula:C₁₂H₁₂F₃N₃
MDL Number:MFCD04968467