Order Entry
Netherlands
ContactUsLinkComponent
1-[4-Cyclopropyl-6-(trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-3-carboxylic acid
1-[4-Cyclopropyl-6-(trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-3-carboxylic acid
Catalog # APOSPC410453-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[4-Cyclopropyl-6-(trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-3-carboxylic acid
Catalog # APOSPC410453-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :298.221 g/mol
Storage Temperature:Ambient
CAS Number:1006448-08-3
Formula:C₁₂H₉F₃N₄O₂
MDL Number:MFCD05861884