Order Entry
Netherlands
ContactUsLinkComponent
1-(4-Fluoro-2-nitrophenyl)-1H-pyrazole-3-carboxylic acid
1-(4-Fluoro-2-nitrophenyl)-1H-pyrazole-3-carboxylic acid
Catalog # APOSPC410441-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(4-Fluoro-2-nitrophenyl)-1H-pyrazole-3-carboxylic acid
Catalog # APOSPC410441-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :251.171 g/mol
Storage Temperature:Ambient
CAS Number:1006442-84-7
Formula:C₁₀H₆FN₃O₄
MDL Number:MFCD08696948