1-(Chloromethyl)-5-methyl-3-(trifluoromethyl)-1H-pyrazole
Catalog # APOSPC410428-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:198.573 g/mol
Storage Temperature:Ambient
CAS Number:1006356-44-0
Formula:C₆H₆ClF₃N₂
MDL Number:MFCD04968631