Order Entry
Netherlands
ContactUsLinkComponent
2-Methyl-3-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
2-Methyl-3-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
Catalog # APOSPC410422-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Methyl-3-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
Catalog # APOSPC410422-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :217.191 g/mol
Storage Temperature:Ambient
CAS Number:1006352-92-6
Formula:C₉H₁₀F₃N₃
MDL Number:MFCD08556255