Order Entry
Netherlands
ContactUsLinkComponent
1-[4-(1,5-Dimethyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidin-2-yl]piperidine-3-carboxylic acid
1-[4-(1,5-Dimethyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidin-2-yl]piperidine-3-carboxylic acid
Catalog # APOSPC410416-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[4-(1,5-Dimethyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidin-2-yl]piperidine-3-carboxylic acid
Catalog # APOSPC410416-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :369.342 g/mol
Storage Temperature:Ambient
CAS Number:1006348-78-2
Formula:C₁₆H₁₈F₃N₅O₂
MDL Number:MFCD05861950