Order Entry
Netherlands
ContactUsLinkComponent
4-Methyl-5-[3-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazol-2-amine
4-Methyl-5-[3-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazol-2-amine
Catalog # APOSPC410412-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Methyl-5-[3-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazol-2-amine
Catalog # APOSPC410412-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :248.228 g/mol
Storage Temperature:Ambient
CAS Number:1006348-68-0
Formula:C₈H₇F₃N₄S
MDL Number:MFCD08558209