Order Entry
Netherlands
ContactUsLinkComponent
 
5-{[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}furan-2-carboxylic acid
Catalog # APOSPC410409-1G
Supplier:  Apollo Scientific
CAS Number:  
5-{[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}furan-2-carboxylic acid
Catalog # APOSPC410409-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :294.614 g/mol
Storage Temperature:Ambient
CAS Number:1006348-61-3
Formula:C₁₀H₆ClF₃N₂O₃
MDL Number:MFCD04970080