Order Entry
Netherlands
ContactUsLinkComponent
2-[(5-{[3,5-Bis(difluoromethyl)-1H-pyrazol-1-yl]methyl}-4-ethyl-4H-1,2,4-triazol-3-yl)sulphanyl]acetohydrazide
2-[(5-{[3,5-Bis(difluoromethyl)-1H-pyrazol-1-yl]methyl}-4-ethyl-4H-1,2,4-triazol-3-yl)sulphanyl]acetohydrazide
Catalog # APOSPC410404-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[(5-{[3,5-Bis(difluoromethyl)-1H-pyrazol-1-yl]methyl}-4-ethyl-4H-1,2,4-triazol-3-yl)sulphanyl]acetohydrazide
Catalog # APOSPC410404-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :381.352 g/mol
Storage Temperature:Ambient
CAS Number:1006343-50-5
Formula:C₁₂H₁₅F₄N₇OS
MDL Number:MFCD03423244