Order Entry
Netherlands
ContactUsLinkComponent
4-Chloro-1-{[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}-1H-pyrazole-3-carboxylic acid
4-Chloro-1-{[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}-1H-pyrazole-3-carboxylic acid
Catalog # APOSPC410401-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-1-{[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}-1H-pyrazole-3-carboxylic acid
Catalog # APOSPC410401-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :308.644 g/mol
Storage Temperature:Ambient
CAS Number:1006341-09-8
Formula:C₁₀H₈ClF₃N₄O₂
MDL Number:MFCD04967777