Order Entry
Netherlands
ContactUsLinkComponent
1,3-Dimethyl-4-nitro-5-(trifluoromethyl)-1H-pyrazole
1,3-Dimethyl-4-nitro-5-(trifluoromethyl)-1H-pyrazole
Catalog # APOSPC410338-1G
Supplier:  Apollo Scientific
CAS Number:  
1,3-Dimethyl-4-nitro-5-(trifluoromethyl)-1H-pyrazole
Catalog # APOSPC410338-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :209.126 g/mol
Storage Temperature:Ambient
CAS Number:1005576-58-8
Formula:C₆H₆F₃N₃O₂
MDL Number:MFCD04970017