Order Entry
Netherlands
ContactUsLinkComponent
2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Catalog # APOSPC410337-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Catalog # APOSPC410337-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :222.164 g/mol
Storage Temperature:Ambient
CAS Number:1005566-34-6
Formula:C₈H₉F₃N₂O₂
MDL Number:MFCD03419687