Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-1-(2-fluorobenzyl)-1H-pyrazol-3-amine
4-Chloro-1-(2-fluorobenzyl)-1H-pyrazol-3-amine
4-Chloro-1-(2-fluorobenzyl)-1H-pyrazol-3-amine
APOSPC410297-1G
:
Apollo Scientific
:
4-Chloro-1-(2-fluorobenzyl)-1H-pyrazol-3-amine
APOSPC410297-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
225.65 g/mol
:
Ambient
:
1001757-51-2
:
C₁₀H₉ClFN₃
:
MFCD02254015